siRNA Details
| Gene Symbol: MGC2889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026877.1 | 1 | 450 | |||
| GenBank mRNA | AK090855.1 | - | 1 | 262 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC2889| Gene Symbol: MGC2889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026877.1 | 1 | 450 | |||
| GenBank mRNA | AK090855.1 | - | 1 | 262 | ||