siRNA Details
| Gene Symbol: MAN1B1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027447.1 | 1 | 841 | |||
| GenBank mRNA | AK023312.1 | - | 1 | 841 | ||
| BC020858.1 | - | 1 | 695 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAN1B1-AS1| Gene Symbol: MAN1B1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027447.1 | 1 | 841 | |||
| GenBank mRNA | AK023312.1 | - | 1 | 841 | ||
| BC020858.1 | - | 1 | 695 | |||