siRNA Details
| Gene Symbol: INE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024616.1 | 1 | 230 | |||
| GenBank mRNA | BC069671.1 | - | 1 | 153 | ||
| BC069696.1 | - | 1 | 152 | |||
| BC069772.1 | - | 1 | 152 | |||
| BC112149.1 | - | 1 | 42 | |||
| BC112175.1 | - | 1 | 42 | |||
| Y10696.1 | - | 1 | 157 | |||

INE1
Genomic Map