siRNA Details
| Gene Symbol: INE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024616.1 | 1 | 175 | |||
| GenBank mRNA | BC069671.1 | - | 1 | 98 | ||
| BC069696.1 | - | 1 | 97 | |||
| BC069772.1 | - | 1 | 97 | |||
| Y10696.1 | - | 1 | 102 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

INE1| Gene Symbol: INE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024616.1 | 1 | 175 | |||
| GenBank mRNA | BC069671.1 | - | 1 | 98 | ||
| BC069696.1 | - | 1 | 97 | |||
| BC069772.1 | - | 1 | 97 | |||
| Y10696.1 | - | 1 | 102 | |||