siRNA Details
| Gene Symbol: LINC00936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028138.1 | 1 | 2149 | |||
| GenBank mRNA | AK094517.1 | - | 1 | 1312 | ||
| BC031240.1 | - | 1 | 1370 | |||
| BC040326.1 | - | 1 | 2143 | |||
| BC047477.1 | - | 1 | 1264 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00936| Gene Symbol: LINC00936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028138.1 | 1 | 2149 | |||
| GenBank mRNA | AK094517.1 | - | 1 | 1312 | ||
| BC031240.1 | - | 1 | 1370 | |||
| BC040326.1 | - | 1 | 2143 | |||
| BC047477.1 | - | 1 | 1264 | |||