siRNA Details
| Gene Symbol: LINC00936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028138.1 | 1 | 2259 | |||
| GenBank mRNA | AK094517.1 | - | 1 | 1422 | ||
| BC031240.1 | - | 1 | 1480 | |||
| BC040326.1 | - | 1 | 2253 | |||
| BC047477.1 | - | 1 | 1374 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00936| Gene Symbol: LINC00936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028138.1 | 1 | 2259 | |||
| GenBank mRNA | AK094517.1 | - | 1 | 1422 | ||
| BC031240.1 | - | 1 | 1480 | |||
| BC040326.1 | - | 1 | 2253 | |||
| BC047477.1 | - | 1 | 1374 | |||