siRNA Details
| Gene Symbol: PERM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291366.1 | NP_001278295.1 | 4 | 3395 | ||
| NM_001291367.1 | NP_001278296.1 | 5 | 3042 | |||
| XM_017002583.1 | XP_016858072.1 | 4 | 3497 | |||
| XM_017002584.1 | XP_016858073.1 | 4 | 3520 | |||
| XM_017002585.1 | XP_016858074.1 | 3 | 3886 | |||
| GenBank mRNA | AK123855.1 | - | 5 | 3018 | ||
| BC006300.2 | - | 2 | 1192 | |||
| F22110.1 | - | 1 | 201 | |||

PERM1
Genomic Map