siRNA Details
| Gene Symbol: PERM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291366.1 | NP_001278295.1 | 3 | 2280 | ||
| NM_001291367.1 | NP_001278296.1 | 4 | 1927 | |||
| XM_017002583.1 | XP_016858072.1 | 3 | 2382 | |||
| XM_017002584.1 | XP_016858073.1 | 3 | 2405 | |||
| GenBank mRNA | AK123855.1 | - | 4 | 1903 | ||
| AK312221.1 | - | 1 | 36 | |||
| BC006300.2 | - | 1 | 77 | |||
| KF150175.1 | - | 3 | 2280 | |||

PERM1
Genomic Map