siRNA Details
Gene Symbol: PERM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291366.1 | NP_001278295.1 | 3 | 2246 | ||
NM_001291367.1 | NP_001278296.1 | 4 | 1893 | |||
XM_017002583.1 | XP_016858072.1 | 3 | 2348 | |||
XM_017002584.1 | XP_016858073.1 | 3 | 2371 | |||
GenBank mRNA | BC006300.2 | - | 1 | 43 | ||
KF150175.1 | - | 3 | 2246 |
