siRNA Details
| Gene Symbol: CROCCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026752.1 | 7 | 1680 | |||
| GenBank mRNA | AK128484.1 | - | 1991 | |||
| BC006312.2 | - | 7 | 1933 | |||
| BC010508.1 | - | 4 | 1205 | |||
| BC047736.1 | - | 7 | 1680 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CROCCP2| Gene Symbol: CROCCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026752.1 | 7 | 1680 | |||
| GenBank mRNA | AK128484.1 | - | 1991 | |||
| BC006312.2 | - | 7 | 1933 | |||
| BC010508.1 | - | 4 | 1205 | |||
| BC047736.1 | - | 7 | 1680 | |||