siRNA Details
| Gene Symbol: CROCCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026752.1 | 7 | 1506 | |||
| GenBank mRNA | AK128484.1 | - | 1817 | |||
| BC006312.2 | - | 7 | 1760 | |||
| BC010508.1 | - | 4 | 1031 | |||
| BC047736.1 | - | 7 | 1506 | |||
| DQ786196.1 | - | 2 | 919 | |||

CROCCP2
Genomic Map