siRNA Details
| Gene Symbol: MGC12916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026880.1 | 1 | 746 | |||
| GenBank mRNA | BC006384.1 | - | 1 | 632 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC12916| Gene Symbol: MGC12916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026880.1 | 1 | 746 | |||
| GenBank mRNA | BC006384.1 | - | 1 | 632 | ||