siRNA Details
| Gene Symbol: ANKRD36BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026844.1 | 1 | 1463 | |||
| GenBank mRNA | BC007072.1 | - | 1 | 1376 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD36BP1| Gene Symbol: ANKRD36BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026844.1 | 1 | 1463 | |||
| GenBank mRNA | BC007072.1 | - | 1 | 1376 | ||