siRNA Details
Gene Symbol: ANKRD36BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026844.1 | 1 | 1646 | |||
GenBank mRNA | BC007072.1 | - | 1 | 1559 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ANKRD36BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026844.1 | 1 | 1646 | |||
GenBank mRNA | BC007072.1 | - | 1 | 1559 |