siRNA Details
| Gene Symbol: PRR34-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027034.1 | 3 | 1559 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR34-AS1| Gene Symbol: PRR34-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027034.1 | 3 | 1559 | |||