siRNA Details
| Gene Symbol: MIR503HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024607.1 | 3 | 314 | |||
| GenBank mRNA | BX090522.1 | - | 3 | 314 | ||
| CN297454.1 | - | 3 | 223 | |||
| KM886853.1 | - | 3 | 195 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR503HG| Gene Symbol: MIR503HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024607.1 | 3 | 314 | |||
| GenBank mRNA | BX090522.1 | - | 3 | 314 | ||
| CN297454.1 | - | 3 | 223 | |||
| KM886853.1 | - | 3 | 195 | |||