siRNA Details
| Gene Symbol: RBM48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032120.3 | NP_115496.2 | 5 | 2820 | ||
| GenBank mRNA | BC007365.2 | - | 1 | 138 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBM48| Gene Symbol: RBM48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032120.3 | NP_115496.2 | 5 | 2820 | ||
| GenBank mRNA | BC007365.2 | - | 1 | 138 | ||