siRNA Details
Gene Symbol: RBM48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032120.3 | NP_115496.2 | 5 | 2985 | ||
GenBank mRNA | BC007365.2 | - | 1 | 303 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RBM48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032120.3 | NP_115496.2 | 5 | 2985 | ||
GenBank mRNA | BC007365.2 | - | 1 | 303 |