siRNA Details
| Gene Symbol: LINC00839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026827.1 | 5 | 1659 | |||
| GenBank mRNA | AK090789.1 | - | 5 | 1606 | ||
| AK092104.1 | - | 5 | 1609 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00839| Gene Symbol: LINC00839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026827.1 | 5 | 1659 | |||
| GenBank mRNA | AK090789.1 | - | 5 | 1606 | ||
| AK092104.1 | - | 5 | 1609 | |||