siRNA Details
| Gene Symbol: HHLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027404.1 | 3, 4 | 446 | |||
| XM_011540548.2 | XP_011538850.1 | 3, 4 | 345 | |||
| GenBank mRNA | AF126164.1 | - | 3, 4 | 427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HHLA3| Gene Symbol: HHLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027404.1 | 3, 4 | 446 | |||
| XM_011540548.2 | XP_011538850.1 | 3, 4 | 345 | |||
| GenBank mRNA | AF126164.1 | - | 3, 4 | 427 | ||