siRNA Details
| Gene Symbol: H2BFXP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003238.2 | 1 | 356 | |||
| Gene Symbol: LOC100101478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130172.1 | 1 | 356 | |||
| GenBank mRNA | AY730279.1 | - | 1 | 356 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ217928.1 | - | 1 | 375 | Chr. X - 104060627 - 104062938 [-] on Build GRCh38 | |

H2BFXP
Genomic Map