siRNA Details
| Gene Symbol: LINC01101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027181.1 | 1 | 231 | |||
| GenBank mRNA | BC112205.1 | - | 1 | 111 | ||
| BC113708.1 | - | 1 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01101| Gene Symbol: LINC01101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027181.1 | 1 | 231 | |||
| GenBank mRNA | BC112205.1 | - | 1 | 111 | ||
| BC113708.1 | - | 1 | 99 | |||