siRNA Details
| Gene Symbol: MUM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024247.1 | 3, 4 | 283 | |||
| XM_011528385.2 | XP_011526687.1 | 4, 5 | 405 | |||
| GenBank mRNA | AK309092.1 | - | 2, 3 | 102 | ||
| AK310721.1 | - | 4, 5 | 317 | |||
| BC130443.1 | - | 3, 4 | 243 | 40146474 | ||
| BC144147.1 | - | 3, 4 | 243 | |||
| U20896.1 | - | 1, 2 | 1212 | |||

MUM1
Genomic Map