siRNA Details
| Gene Symbol: HBBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001589.1 | 3 | 621 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | U01317.3 | - | 3 | 47229 | Chr. 11 - 5215910 - 5289430 [-] on Build GRCh38 | |

HBBP1
Genomic Map