siRNA Details
| Gene Symbol: TINCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027064.2 | 3 | 3673 | |||
| GenBank mRNA | BC036545.1 | - | 3 | 3638 | ||
| BC047562.1 | - | 1 | 1156 | |||
| Gene Symbol: CARMIL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125696.1 | - | 1837 | |||

TINCR
Genomic Map