siRNA Details
| Gene Symbol: TINCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027064.2 | 3 | 2781 | |||
| GenBank mRNA | BC036545.1 | - | 3 | 2746 | ||
| BC047562.1 | - | 1 | 264 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG354568.1 | - | 1 | 176 | Chr. 19 - 5558175 - 5559294 [-] on Build GRCh38 | |

TINCR
Genomic Map