siRNA Details
| Gene Symbol: SNHG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003672.2 | 2 | 1018 | |||
| NR_024542.1 | 3 | 389 | ||||
| NR_024543.1 | 3 | 389 | ||||
| GenBank mRNA | AK090585.1 | - | 2 | 1013 | ||
| BC042369.1 | - | 3 | 409 | |||
| BC042667.1 | - | 2 | 1032 | |||
| Gene Symbol: SNORA17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054908.1 | - | 3 | 389 | ||

SNHG7
Genomic Map