siRNA Details
| Gene Symbol: MIR22HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028502.1 | 2 | 2182 | |||
| NR_028503.1 | 3 | 1375 | ||||
| NR_028504.1 | 4 | 962 | ||||
| NR_028505.1 | 3 | 879 | ||||
| GenBank mRNA | AF070569.1 | - | 1 | 1751 | ||
| BX640650.1 | - | 4 | 900 | |||
| BX648321.1 | - | 2 | 2119 | |||

MIR22HG
Genomic Map