siRNA Details
| Gene Symbol: TISP43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027313.1 | 3 | 1565 | |||
| NR_027314.1 | 2 | 1389 | ||||
| GenBank mRNA | BC037249.1 | - | 2 | 1390 | ||
| BC064385.1 | - | 3 | 1565 | |||
| Gene Symbol: LOC646743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033930.1 | 3 | 1582 | |||

TISP43
Genomic Map