siRNA Details
| Gene Symbol: URB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026845.1 | 1 | 539 | |||
| GenBank mRNA | AK026376.1 | - | 1 | 517 | ||
| BC007928.1 | - | 1 | 545 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

URB1-AS1| Gene Symbol: URB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026845.1 | 1 | 539 | |||
| GenBank mRNA | AK026376.1 | - | 1 | 517 | ||
| BC007928.1 | - | 1 | 545 | |||