siRNA Details
| Gene Symbol: URB1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026845.1 | 1 | 306 | |||
| GenBank mRNA | AK026376.1 | - | 1 | 284 | ||
| BC007928.1 | - | 1 | 312 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI756790.1 | - | 1 | 281 | Chr. 21 - 32393158 - 32393962 [+] on Build GRCh38 | |

URB1-AS1
Genomic Map