siRNA Details
| Gene Symbol: MGC16275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026914.1 | 2 | 1335 | |||
| GenBank mRNA | BC043607.1 | - | 2 | 1335 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC16275| Gene Symbol: MGC16275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026914.1 | 2 | 1335 | |||
| GenBank mRNA | BC043607.1 | - | 2 | 1335 | ||