siRNA Details
| Gene Symbol: WHAMMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003521.1 | 5 | 1195 | |||
| GenBank mRNA | AL833132.1 | - | 4 | 647 | ||
| BC018097.1 | - | 4 | 514 | |||
| BC044610.1 | - | 4 | 647 | |||
| BC048987.1 | - | 5 | 1255 | |||
| BC093703.1 | - | 2 | 252 | |||
| BC093705.1 | - | 2 | 252 | |||
| DQ309039.1 | - | 4 | 504 | |||
| DQ309040.1 | - | 5 | 1195 | |||

WHAMMP3
Genomic Map