siRNA Details
| Gene Symbol: LINC00479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027272.1 | 6 | 656 | |||
| GenBank mRNA | AK057397.1 | - | 6 | 656 | ||
| BC128197.1 | - | 2 | 251 | |||
| BC128198.1 | - | 6 | 677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ENSG00000236384
LINC00479| Gene Symbol: LINC00479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027272.1 | 6 | 656 | |||
| GenBank mRNA | AK057397.1 | - | 6 | 656 | ||
| BC128197.1 | - | 2 | 251 | |||
| BC128198.1 | - | 6 | 677 | |||