siRNA Details
| Gene Symbol: SPATA41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028139.1 | 2 | 2259 | |||
| NR_028140.1 | 3 | 667 | ||||
| GenBank mRNA | AK057536.1 | - | 2 | 1196 | ||
| BC028144.1 | - | 3 | 663 | 5214282 | ||
| BC132704.1 | - | 3 | 610 | |||
| BC132706.1 | - | 3 | 610 | |||

SPATA41
Genomic Map