siRNA Details
| Gene Symbol: SPATA41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028139.1 | 2 | 2767 | |||
| NR_028140.1 | 3 | 1175 | ||||
| GenBank mRNA | AK057536.1 | - | 2 | 1704 | ||
| BC028144.1 | - | 3 | 1171 | 5214282 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000560282.1 | - | 2 | 1704 | Chr. 15 - 100344478 - 100349655 [-] on Build GRCh38 | |

SPATA41
Genomic Map