siRNA Details
| Gene Symbol: SPATA41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028139.1 | 1 | 511 | |||
| NR_028140.1 | 1 | 26 | ||||
| GenBank mRNA | AK124283.1 | - | 1 | 506 | ||
| BC028144.1 | - | 1 | 22 | 5214282 | ||
| DB060272.1 | - | 1 | 513 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA41| Gene Symbol: SPATA41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028139.1 | 1 | 511 | |||
| NR_028140.1 | 1 | 26 | ||||
| GenBank mRNA | AK124283.1 | - | 1 | 506 | ||
| BC028144.1 | - | 1 | 22 | 5214282 | ||
| DB060272.1 | - | 1 | 513 | |||