siRNA Details
| Gene Symbol: SPATA41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028139.1 | 2 | 2570 | |||
| NR_028140.1 | 3 | 978 | ||||
| GenBank mRNA | AK057536.1 | - | 2 | 1507 | ||
| BC028144.1 | - | 3 | 974 | 5214282 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA41| Gene Symbol: SPATA41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028139.1 | 2 | 2570 | |||
| NR_028140.1 | 3 | 978 | ||||
| GenBank mRNA | AK057536.1 | - | 2 | 1507 | ||
| BC028144.1 | - | 3 | 974 | 5214282 | ||