siRNA Details
| Gene Symbol: TRIM78P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF230412.1 | - | LIT2077 TRIMP pseudogene |
2 | 583 | |
| AL360159.1 | - | 2 | 583 | |||
| AL360183.1 | - | 1 | 331 | |||

TRIM78P
Genomic Map