siRNA Details
| Gene Symbol: TDRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024015.1 | 11 | ||||
| GenBank mRNA | BC033995.2 | - | 1 | 98 | ||
| BC042123.1 | - | 1 | 98 | |||
| DQ168992.1 | - | 11 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TDRG1| Gene Symbol: TDRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024015.1 | 11 | ||||
| GenBank mRNA | BC033995.2 | - | 1 | 98 | ||
| BC042123.1 | - | 1 | 98 | |||
| DQ168992.1 | - | 11 | ||||