siRNA Details
| Gene Symbol: TDRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024015.1 | 2 | 964 | |||
| GenBank mRNA | BC033995.2 | - | 2 | 1051 | ||
| BC042123.1 | - | 2 | 1051 | |||
| DQ168992.1 | - | 2 | 964 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TDRG1| Gene Symbol: TDRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024015.1 | 2 | 964 | |||
| GenBank mRNA | BC033995.2 | - | 2 | 1051 | ||
| BC042123.1 | - | 2 | 1051 | |||
| DQ168992.1 | - | 2 | 964 | |||