siRNA Details
| Gene Symbol: SMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024054.2 | 6, 7 | 782 | |||
| GenBank mRNA | X75940.1 | - | 6, 7 | 783 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMA4| Gene Symbol: SMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024054.2 | 6, 7 | 782 | |||
| GenBank mRNA | X75940.1 | - | 6, 7 | 783 | ||