siRNA Details
| Gene Symbol: SMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024054.2 | 4, 5 | 588 | |||
| GenBank mRNA | X75940.1 | - | 4, 5 | 589 | ||
| Gene Symbol: SMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK289851.1 | - | 7, 8 | 882 | ||

SMA4
Genomic Map