siRNA Details
Gene Symbol: LINC00917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024406.1 | 6 | 1730 | |||
GenBank mRNA | AK093244.1 | - | 6 | 1730 | ||
BC043250.1 | - | 5 | 1787 | |||
BC144028.1 | - | 4 | 950 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL137562.1 | - | 4 | 638 | Chr. 16 - 86331844 - 86337556 [-] on Build GRCh38 |
