siRNA Details
Gene Symbol: HAVCR1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003603.1 | 1 | 404 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HAVCR1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003603.1 | 1 | 404 |