siRNA Details
| Gene Symbol: DCAF13P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027642.1 | 1 | 123 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCAF13P3| Gene Symbol: DCAF13P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027642.1 | 1 | 123 | |||