siRNA Details
| Gene Symbol: LINC00704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024475.1 | 2 | 154 | |||
| GenBank mRNA | BC048320.1 | - | 2 | 154 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00704| Gene Symbol: LINC00704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024475.1 | 2 | 154 | |||
| GenBank mRNA | BC048320.1 | - | 2 | 154 | ||