siRNA Details
| Gene Symbol: ZNF876P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027481.1 | 2 | 600 | |||
| GenBank mRNA | BC028359.1 | - | 2 | 595 | ||
| BC046475.1 | - | 2 | 571 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI463333.1 | - | 2 | 576 | Chr. 4 - 212629 - 254157 [+] on Build GRCh38 | |

ZNF876P
Genomic Map