siRNA Details
Gene Symbol: HSPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024151.1 | 1 | 749 | |||
GenBank mRNA | CA391777.1 | - | 1 | 60 | ||
DR004990.1 | - | 1 | 695 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HSPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024151.1 | 1 | 749 | |||
GenBank mRNA | CA391777.1 | - | 1 | 60 | ||
DR004990.1 | - | 1 | 695 |