siRNA Details
| Gene Symbol: LOC93429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027003.1 | 6 | 679 | |||
| GenBank mRNA | AL137376.1 | - | 6 | 678 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC93429| Gene Symbol: LOC93429 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027003.1 | 6 | 679 | |||
| GenBank mRNA | AL137376.1 | - | 6 | 678 | ||